Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA V-Gard H2™ Replacement Sweatband, MIPS

Catalog No. 17200686 Shop All MSA Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-686 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-686 Supplier MSA Supplier No. 10246731
Only null left
Add to Cart
Add to Cart

Replacement sweatband assembly for V-Gard H2™ Safety Helmet with MIPS. Antimicrobial treatment with Ionic+ high purity silver.

  • Accessory

Specifications

Product Type Sweatband
Size Universal
Material Cotton Terrycloth
Color Black
Closure Type Snap
Disposable Reusable
For Use With (Application) Construction, Oil & Gas, Utilities, Mining, Manufacturing, Confined Space, Safety at Heights,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.