Learn More
MSA V-Gard H2™ Safety Helmet, MIPS, Non-Vented
Catalog No. 17200689 Shop All MSA Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-200-689 White
1 options
Catalog No. 17-200-689 Supplier MSA Supplier No. 10242822
Add to Cart
Add to Cart
ANSI Z89.1 Type II certification with integrated chin strap. MIPS Safety System for rotational impact protection.
The V-Gard H2™ Safety Helmet provides superior comfort and versatility while incorporating the latest technology to help protect against lateral impacts. The optional MIPS™ brain protection system for industrial safety helmets adds another layer of protection designed to help reduce the risk of brain trauma by redirecting rotational energies in the event of certain angled impacts.
  • ANSI Z89.1 Type II certification with integrated chin strap for enhanced protection against lateral impacts, including during slips, trips and falls
  • MIPS™ brain protection system offers additional rotational impact protection to help reduce the risk of traumatic brain injury resulting from slips, trips and falls
  • Extensive accessory compatibility with nearly all V-Gard H1 and V-Gard accessories for a wide variety of industrial applications and easy integration with existing inventory
  • Quick and easy customization with thousands of existing logos, easy creation of new logos, and reflective stripes
  • Comfortable ratchet suspension that's easily adjustable with a gloved hand
  • Antimicrobial treatment of sweatband, crown pad and ratchet pad with Ionic+™ high purity silver to eliminate odor-causing bacteria
  • Integrated headlamp strap clips for easy and secure donning and doffing of lights on the helmet
Specifications
Color White
Product Type Climbing Helmet
Size 6.5 to 8
Ventilated Not Vented
Head Style Front Brim
Width (Metric) 8.54 in.
Width (English) 8.54 in.