Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA V-Gard H2™ Safety Helmet, Vented

Catalog No. 17200699 Shop All MSA Products
Change view
Click to view available options
Size:
6.5 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-699 6.5 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-699 Supplier MSA Supplier No. 10242629
Add to Cart
Add to Cart

ANSI Z89.1 Type II certification with integrated chin strap. MIPS Safety System for rotational impact protection.

The V-Gard H2™ Safety Helmet provides superior comfort and versatility while incorporating the latest technology to help protect against lateral impacts. The optional MIPS™ brain protection system for industrial safety helmets adds another layer of protection designed to help reduce the risk of brain trauma by redirecting rotational energies in the event of certain angled impacts.

  • ANSI Z89.1 Type II certification with integrated chin strap for enhanced protection against lateral impacts, including during slips, trips and falls
  • Venting for improved air flow, cooling and reduced heat stress. Non-vented options for use in electrical applications where Class E certification is needed
  • Extensive accessory compatibility with nearly all V-Gard H1 and V-Gard accessories for a wide variety of industrial applications and easy integration with existing inventory
  • Quick and easy customization with thousands of existing logos, easy creation of new logos, and reflective stripes
  • Comfortable ratchet suspension that's easily adjustable with a gloved hand
  • Antimicrobial treatment of sweatband, crown pad and ratchet pad with Ionic+™ high purity silver to eliminate odor-causing bacteria
  • Integrated headlamp strap clips for easy and secure donning and doffing of lights on the helmet

Specifications

Color White
For Use With (Equipment) Hard Hats
Product Type Climbing Helmet
Size 6.5 to 8
Ventilated Vented
Head Style Front Brim
Width (Metric) 8.54 in.
Width (English) 8.54 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.