Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-Gard™ HDPE Frame for Universal MSA™ Hats without Debris Control

Catalog No. 19141450 Shop All MSA Products
Change view
Click to view available options
Material:
HDPE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-141-450 HDPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-141-450 Supplier MSA™ Supplier No. 10116628
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a universal frame with a heavy-duty, flat, rubber strap that can be used with slotted and non-slotted MSA™ helmets. MSA™ V-Gard™ HDPE Frame for Universal MSA™ Hats without Debris Control has a unique sloped frame design so material slides off the frame and out of site.

  • Unique 3-point visor loading to fit and remove visors quickly
  • Sloped design allows material to easily slide off the frame
  • Works with or without ear muffs
  • Perfect Compatibility with many half-mask respirators and more
  • Non-metal construction for use in electrical applications
  • Replaceable, optional debris control creates tight seal between frame and helmet
  • Universal Heavy-duty, flat, rubber strap for easy and secure placement on MSA slotted and non-slotted  helmets
  • ET frames are tested to temperatures of 176.7°C, reducing warping, cracking or crazing.

Specifications

Color Black
For Use With (Equipment) MSA Universal Caps
Material HDPE
Product Type Visor Frame
Quantity 1 Ea.
Suspension Type 3 point
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.