Learn More
MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Catalog No. 19815529 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Orange
Orange
Pink
Purple
Red
White
Yellow
Size:
Standard
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-815-529 Pink Standard
19-062899 Orange Standard
19-080144 Hi-vis Orange Standard
19-058270 Green Standard
18-999-3987 Purple Standard
18-999-3985 Black Standard
19-054727 White Standard
19-054725 Blue Standard
19-054728 Yellow Standard
19-054726 Red Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-815-529 Supplier MSA™ Supplier No. 485364
Add to Cart
Add to Cart
Lightweight for all-day wearing comfort
  • Four-point webbed Staz-On suspension with perforated vinyl sweatband
  • Sizes: Small (6-7 1/8), Standard (6 1/2-8), and Large (7-8 1/2)
  • Available in more than 20 colors, with custom imprinting and striping available
Order Info
Contact your Fisher Safety Customer Safety Representative for ordering information for sizes and colors not shown.
†With lamp bracket and cord holder (Class C)
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Color Pink
Size Standard
Suspension Type Staz-On Pin-Lock
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.