Learn More
MSA™ V-Gard™ Visor
V-Gard polycarbonate shaded visors are thick and molded to protect against damage from impact, elevated temperatures, weld spatter and infrared (IR) radiation. Ideal for autogenous welding, metal pouring, gas soldering and light cutting/brazing, they’re engineered for heavy-duty use.
$39.75
Specifications
Lens Coating Anti-scratch, Anti-fog, Impact-resistant
Product Type Visor
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-030-078
View Documents
MSA™
10246714
IR Shade 10
Each for $39.75
Only null left
Add to Cart
19-030-079
View Documents
MSA™
10246715
IR Shade 12
Each for $39.75
Only null left
Add to Cart
Description
Description
  • Thickness helps prevent pitting and warping from heat or weld spatter
  • Suitable for applications where welding, cutting and brazing hazards exist
  • V-Gard polycarbonate shaded visors are thick and molded to protect against damage from impact, elevated temperatures, weld spatter and infrared (IR) radiation
  • Ideal for autogenous welding, metal pouring, gas soldering and light cutting/brazing, they are engineered for heavy-duty use
Specifications
Specifications
Anti-scratch, Anti-fog, Impact-resistant
Polycarbonate
Polycarbonate
None
Visor
Reusable
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.