Learn More
Leica Biosystems v-TRACT CARTRIDGE + 3 BACTIGUA
Supplier:  Leica Biosystems 3804954
Encompass
The X-Tract Filtration System is not available for purchase. However, the X-Tract Filters and Bactiguards listed below are available.The accessories for the X-Tract Filtration system are for removing hazardous vapours from localized sources. It will remove both formalin and xylene vapours while its Bactiguard Pre-Filter will eliminate microbes and viruses.The unit draws contaminated air through a sealed replaceable cartridge containing material which attracts, reacts with and removes vapour molecules.
Catalog No. 50-255-2422
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.