Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Tocopheryl acetate (Vitamin E acetate) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco V0331ML
Description
Tocopheryl acetate is a natural form of Vitamin E, used as a fortifying agent and dietary supplement for Vitamin E in food. It is also the main ingredient of skin care products and dermatological preparations. Tocopheryl acetate in combination with α-tocopherol has been used in the therapy of conditions such as sterility, muscular dystrophy, cardiac vascular disorders and anemia.Specifications
75-05-8, 52225-20-4 | |
UN 1648 3 / PGII | |
472.74 | |
1 mL |
C31H52O3 | |
1.0 mg/mL in methanol | |
Ampule |
Safety and Handling
EINECSNumber : 200-835-2
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction