Learn More
PIP™ v2™ V2 Passive Ear Muff with Adjustable Headband - NRR 25
Catalog No. 17489668 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
25
Catalog No. Noise Reduction Rating (NRR)
17-489-668 25
1 options
Catalog No. 17-489-668 Supplier PIP™ Supplier No. 263V2HB
  • $37.50
    $34.65 / Pair of 1
    Save $2.85
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
V2 oassive ear muff with adjustable headband - NRR 25.
  • Dielectric, metal-free earmuffs suitable for use in workplace environments with exposure to electrical voltage
  • Smooth telescopic size adjustment for precise and stable fit
  • Slim stirrup design reduces the risk of snagging in confined areas
  • Extra soft memory foam ear cushions and ultra low weight padded headband provide optimum comfort even when worn for extended periods
  • Bright Color coding with v1™,v2™,v3™ noise protection indicator for easy selection and identification
  • Replacement hygiene kits available separately
Specifications
Noise Reduction Rating (NRR) 25
Corded No
Product Type Headband Ear Muff
Disposable Reusable
Metal Detectable No
Headband Color Black
Dielectric Yes
Style Over-the-head
Foldable No