Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ V4500 Barcode Scanner USB Cable

Catalog No. 17986741 Shop All Brady Corporation Products
Change view
Click to view available options
Length (English):
3.28 ft.
9.19 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-986-741 9.19 ft.
17-986-742 3.28 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-986-741 Supplier Brady™ Supplier No. 177238
Only null left
Add to Cart
Add to Cart

V4500 Barcode Scanner accessories to ensure uninterrupted productivity.

Cable - USB Cable for V4500 Barcode Scanner

Specifications

Description USB to RJ50-28 Cable for V4500 Barcode Reader
Weight (Metric) 0.13 kg
Weight (English) 0.287 lb.
For Use With (Application) Asset Tracking, General Identification, Barcode Labelling, Circuit Board Labelling, Component and Equipment Labelling, Food and Beverage Process Labelling, Inventory and Inspection Labelling
For Use With (Equipment) V1300 Cradle (177235), V1300 Cradle Kit (177232), V4500 Barcode Scanner Kit (177229)
Length (English) 9.19 ft.
Type Cable
Alternate Part Number BRA-USB-RJ50-28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.