Learn More
Sino Biological V5 Synthetic Peptide 5mg
Supplier:  Sino Biological PP1003785
Encompass_Preferred
V5 peptide can be used to competitively elute V5-tagged fusion proteins from immobilized anti-V5 magnetic beads. Use V5 peptide to elute V5-tagged proteins.
Catalog No. NC1597451
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.