Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V5 Tag Monoclonal Antibody (SV5-Pk1), FITC
GREENER_CHOICE

Catalog No. PIMA180281
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180281 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180281 Supplier Invitrogen™ Supplier No. MA180281
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-80281 recognizes the sequence IPNPLLGL, present on the P/V proteins of the paramyxovirus, SV5. This antibody has been used to detect recombinant proteins, including epitope-tagged transmembrane and secreted proteins. Successfully used tags include a 14-amino acid tag (gly lys pro ile pro asn pro leu leu gly leu asp ser thr) and a smaller 9-amino acid tag that has been used to inhibit the binding of the monoclonal antibody to its native protein.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen V5 Tag
Applications Immunocytochemistry
Classification Monoclonal
Clone SV5-Pk1
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Alias simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
Host Species Mouse
Immunogen Paramyxovirus SV5, Simian-Virus 5.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.