Learn More
Leybold Vacuum Products Inc Leybonol LVO 100 Mineral Oil Free of Additives, 208 L
Supplier:  Leybold Vacuum Products Inc L10099
Encompass_Preferred
Mineral oil, free of additives. Low vapor pressure, minimal foaming, very good water separation. Standard oil for low ultimate pressures. For pumping air, chemically inert gases and water vapor.
Catalog No. NC1679422
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.