Learn More
Med Vet International Furosemide Tablets 12.5mg, 500 Count
Supplier: Med Vet International RXVFUROSE12.5
Furosemide Tablets are an effective diuretic-saluretic for oral use in the treatment of edema (pulmonary congestion, ascites) associated with cardiac insufficiency and acute noninflammatory tissue edema in dogs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.