Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vaccinia Virus B18R (Carrier-Free) Recombinant Protein, eBioscience™

Catalog No. 50245764
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-764 50 μg
34-818-586 1 mg
50-933-2 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-245-764 Supplier Invitrogen™ Supplier No. 34818581
Add to Cart
Add to Cart

Recombinant Protein

The B18R protein is a vaccinia virus-encoded receptor with specificity for mouse, human, rabbit, pig, rat, and cow type I interferons which has potent neutralizing activity.B18R that acts as a decoy receptor for Type I Interferons (IFNa, IFNb, IFNe,k,t,d,z, w,v). B18R was recently identified to enable increased cell viability during RNA transfection protocols designed to convert human somatic donor cells into iPSCs via direct delivery of modified synthetic mRNAs for OCT4, SOX2, KLF4 and MYC (OSKM) and Lin28 with the aim to enable highly efficient reprogramming of somatic cells to pluripotency. This allows for re-directed differentiation toward a desired lineage while removing the risk of genomic integration and insertional mutagenesis inherent to DNA-bases methodologies and eliminates the need for virus-based approaches. iPSCs represent a widely available, non-controversial and practically infinite source of pluripotent cells.The B18R protein is a type I interferon receptor encoded by the B18R gene of the Western Reserve vaccinia virus strain. The 60-65 kD glycoprotein is related to the interleukin-1 receptors and is a member of the immunoglobulin superfamily, unlike other type I IFN-receptors, which belong to the class II cytokine receptor family. The B18R protein has a high affinity (KD, 174 pM) for human IFN alpha and, unlike other type I IFN receptors, has broad species specificity, binding to type I interferons of human, mouse, rat, rabbit, pig, and cow. Among viral host response modifiers, the B18R protein is unique in that it exists as a soluble extracellular, as well as a cell surface protein, enabling blockage of both autocrine and paracrine IFN functions. The B18R protein has been shown to inhibit the antiviral potency of IFN-alpha1, IFN-alpha2, IFN-alpha-8/1/8, and IFN-omega on human cells. The soluble B18R protein is highly potent for neutralizing type I interferons, which include IFN-alpha, beta, delta, kappa. Please note effects on mouse IFNs vary from other species; it has been shown that B18R does not neutralize mouse IFN beta.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) Bioactivity, Functional Assay
Formulation PBS with no preservative; pH 7.5
Molecular Weight (g/mol) 48 kDa
Name Vaccinia Virus B18R (Carrier-Free)
pH Range 7.5
Purification Method Purified
Quantity 50 μg
Source Vaccinia Virus
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.01 ng/μg
Common Name Vaccinia Virus B18R
Gene Symbol Vaccinia Virus-Encoded Neutralizing Type I Interferon Receptor; Type I IFN inhibitor
Biological Activity Pre-incubation of recombinant IFN-alpha 2 A with 0.1 μg/mL B18R for 1 hour (at RT was found to completely neutralize the activity of 1 ng/mL IFN-alpha 2 a.)
Product Type Protein
Conjugate Unconjugated
Species Bovine, Human, Mouse, Pig, Rabbit, Rat
Recombinant Recombinant
Protein Tag His-tag
Sequence Vaccinia Virus B18R, ammino acids His20-Glu351 (Accession # D01019) with a His-tag
Content And Storage -80°C
Activity Pre-incubation of recombinant IFN-alpha 2a with 0.1 μg/mL B18R for 1 hour at RT was found to completely neutralize the activity of 1 ng/mL IFN-alpha 2a.
Expression System Insect cells
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.