Learn More
Cole-Parmer Whatman 6722-5000 50 Filter, 0.45 mm PTFE, 50 mm Dia; 10/Pk
Supplier:  Cole-Parmer 2970100
Encompass_Preferred
Use this filter to prolong the life of your vacuum pump and reduce pump maintenance problems. Encapsulated in a polypropylene (PP) housing, the fully hydrophobic PTFE membrane prevents liquid and aerosol contamination of your pump. The membrane retains 99.99% of airbone particles greater than 0.1 µm. Max flow: 65 sLpm @ 14 psi.
Catalog No. NC9735924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.