Learn More
CHEMetrics Vacu-vials Test Kit, Ammonia 2

Description
Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater
- Each vacuum-sealed ampule contains a unit dose of premeasured reagent
- Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
- Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
- Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
- For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units
Specifications
Specifications
| Analytes | Ammonia 2 |
| For Use With (Equipment) | V-2000 photometer or spectrophotometer |
| Includes | Cardboard box, 30 ampoules, stabilizer solution, 25 mL sample cup, ampoule blank, instructions |
| No. of Tests | 30 |
| Range | 0 to 14 ppm |
| Test Method | Direct nesslerization |
| Test Time | 2 min. |
| Type | Vacu-vials Instrumental Test Kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.