Learn More
Integra Biosciences Corp INTEGRA VACUBOY, Vacuum Hand Operator
SDP
Supplier:  Integra Biosciences Corp 155510
Encompass
VACUBOY hand operator with 1-Channel stainless aspiration adapter 40 mm and 1-Channel plastic aspiration adapter for disposable tips with ejector, incl. rubber adapter for Pasteur Pipettes.
Catalog No. NC0150162