Learn More
Cytiva VacuCap 90 Vacuum Filtration Devices - 0.45 µm, 60 mm, gamma-irradiated (10/pkg)
Supplier:  Cytiva 4634
Encompass_Preferred
Innovative Bottle-Top Filters for Fast Vacuum Filtration of 100 mL up to 5 L of Aqueous Solutions, Vacuum Filtration Made Easy-Replace current vacuum filter systems, simplify workflow, maximize filtration-Supor membrane provides high flow rates.-No need to constantly refill upper fluid reservoir.-Draws directly from the mixing reservoir.-Eliminates the possibility of contamination from transfer steps. Filters directly into the desired container.-Available built-in prefilter increases throughput of particulate-laden solutions such as serum-containing media.-Environmentally-friendly with minimal plastic waste and smaller storage footprint.-Patented small design accepts a variety of collection vessels.-TA version available with tubing attached to each device for maximum convenience.-Gamma irradiated, eliminates potential contamination by EtO residuals. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9602
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.