Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One VACUETTE™ EVOPROTECT SAFETY Blood Collection/Infusion Set

Catalog No. 22405166
Change view
Click to view available options
Needle Gauge:
21 G
23 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-405-166 21 G
22-405-167 23 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-405-166 Supplier Greiner Bio-One Supplier No. 450120
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blood collection/infusion set is not made with natural rubber latex or DEHP

  • Blood collection/infusion set
  • Single packed
  • Sterile
  • 0.75 in. tubing
TRUSTED_SUSTAINABILITY

Specifications

Product Type Safety Blood Collection/Infusion Set
Color Green
Needle Gauge 21 G
Sterility Sterile
For Use With (Application) Blood collection/infusion
Length (English) Tube 7.5 in.
Quantity 60 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.