Learn More
Eppendorf™ Vacufuge Plus - Centrifuge Concentrator
Catalog No. 05414174 Shop All Eppendorf North America Products
Change view
Click to view available options
Pump Type:
Integrated Diaphragm Vacuum Pump
Integrated Diaphragm Vacuum Pump w/Connection
Without pump
Includes:
Rotor F-45-48-11
Without Rotor
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pump Type Includes
05-414-174 Integrated Diaphragm Vacuum Pump Without Rotor
07-748-13 Without pump Rotor F-45-48-11
05-414-177 Integrated Diaphragm Vacuum Pump w/Connection Without Rotor
07-748-15 Integrated Diaphragm Vacuum Pump Rotor F-45-48-11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 05-414-174 Supplier Eppendorf™ Supplier No. 022820168
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The Eppendorf Vacufuge Plus facilitates fast and efficient, gentle vacuum concentration of DNA, RNA, nucleotides, proteins and other liquid or wet samples with its advanced heating technology. Its coated lid provides superior chemical resistance against many acids and organic solvents.
  • Comes in 3 system configurations
  • Extremely quiet operation <50 dB(A)
  • Chemical-resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
  • Small footprint saves valuable bench space
  • Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
  • Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
  • Can be connected with most commercially available cold traps for use with vapors
  • Coated lid with superior chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Model Vacufuge Plus
Includes Without Rotor
Pump Type Integrated Diaphragm Vacuum Pump
Voltage 120 V
Type Centrifuge Concentrator