Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Polyanethol Sulfonate Additive, 8.3 mL, Conventional Closure Glass Tube, 100/Box
Supplier:  Med Vet International 364960
Encompass_Preferred
Features Additive: Sodium polyanethol sulfonate 0.35% in 0.85% sodium chloride, 1.7mL With Transparent Tube Label
Catalog No. 50-304-4590