Description
WHY IT'S NEEDED
Labile metabolic peptides (e.g., GLP-1, GIP, glucagon, OXM, ghrelin) degrade rapidly in conventional EDTA plasma, reducing accuracy and reproducibility. Stabilizing intact analytes early minimizes preanalytical variability.
HOW IT WORKS
A standardized workflow and a specialized tube with proprietary protease, esterase, and DPP-IV inhibitors that act immediately at blood draw to halt enzymatic activity
Specifications
Specifications
| Color | Clear |
| Product Type | Blood Collection Tube |
| Additive | None |
| Material | Plastic |
| Sterility | Sterile |
| Closure Type | Hemogard |
| Capacity (Metric) | 2 mL |