Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture PROMO

Catalog No. 13688012PM Shop All BrandTech Scientific Products
Change view
Click to view available options
Includes:
4 L PP Bottle, External Liquid Level Sensor
4 L PP Bottle, Vacuum Pump
Overall Height (English):
19.68 in.
Weight (Metric):
7.3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Overall Height (English) Weight (Metric)
13-688-012PM 4 L PP Bottle, External Liquid Level Sensor 19.68 in. 7.3 kg
13-688-008PM 4 L PP Bottle, Vacuum Pump 19.68 in. 7.3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-688-012PM Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20727403PM

BVC Fluid Aspiration Systems for Cell Culture are designed for optimal vacuum control, efficient supernatant aspiration, and safe disposal of biological fluids.

Three models to choose from - basic, control, and professional

  • Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
  • G versions feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
  • Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
  • The VacuuHandControl (VHC) keeps all fluid within aspiration tubing ensuring ultimate containment and cleaning convenience
  • The control and professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
  • Streamlined design incorporates bottle, controls and pump (control and professional models), in a lab friendly small footprint
  • Quick connectors are supplied with the standard (non-G) professional, but can be added to the other five models

BVC basic

  • The unpowered version, suitable for use with a VACUULAN™ vacuum network, central vacuum system or shared pump
  • Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
  • Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl

BVC control

  • The BVC control features a powerful whisper-quiet VACUUBRAND™ chemistry-design vacuum pump, eliminating the need for central vacuum supply
  • Potentially biohazardous materials can be contained in a local area
  • The pump is turned on and off automatically on demand when using the VacuuHandControl, which further reduces the noise level
  • An easy to clean touch panel adjusts and indicates the vacuum level switchpoint

BVC professional

  • Includes an external liquid level sensor, which operates without contacting bottle contents
  • Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
  • The standard version is supplied with quick connectors
  • The G version includes hose barb connections; optional quick connectors are available

Available Accessories:

  • Replacement filter: 20638266
  • Replacement VHCpro tubing (sold per meter, minimum order of 2 meters): 20636156
  • VHCpro hand controller (one supplied with unit): 20688061
  • VHCpro stand: 20635680
  • Pipette tip ejector for VHCpro: 20635638
  • 8-channel adapter with tip ejector for VHCpro: 20635679
  • BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC): 20696880
  • Quick connect for second user (between VHCpro and bottle): 20635807
  • Standard connect for second user: 20699943
  • Quick connect (between bottle and pump): 20635808
  • Replacement 4 L PP bottle (with cap): 20635810
  • Replacement 2 L glass bottle (with cap): 20635809
  • 2 L glass bottle adapter: 20635839
TRUSTED_SUSTAINABILITY

Specifications

Max. Vacuum 150 mbar (112 Torr)
Control Touchpanel
Shipping Weight (English) 17 lb.
Shipping Weight (Metric) 7.7 kg
Includes 4 L PP Bottle, External Liquid Level Sensor
Electrical Requirements 100/120 V, 50/60 Hz
Model BVC Professional
Noise Level 45 dBA
Plug Type US
Temperature (Metric) Storage -10°C to +60°C
Weight (English) 16.09 lb.
Weight (Metric) 7.3 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 0.4 to 0.5 CFM
Frequency 50/60 Hz
Overall Height (English) 19.68 in.
Overall Height (Metric) 50 cm
Overall Length (English) 16.06 in.
Overall Length (Metric) 40.8 cm
Overall Width (English) 7.63 in.
Pump Type Chemistry Diaphragm Pump
Enclosure Rating IP40
Maximum Pressure 1.1 bar
Voltage 100/120 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.