Learn More
BRANDTECH™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture PROMO
BVC Fluid Aspiration Systems for Cell Culture are designed for optimal vacuum control, efficient supernatant aspiration, and safe disposal of biological fluids.
Supplier: BRANDTECH™ VACUUBRAND™ 20727503PM
Description
Three models to choose from - basic, control, and professional
- Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
- G versions feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
- Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
- The VacuuHandControl (VHC) keeps all fluid within aspiration tubing ensuring ultimate containment and cleaning convenience
- The control and professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
- Streamlined design incorporates bottle, controls and pump (control and professional models), in a lab friendly small footprint
- Quick connectors are supplied with the standard (non-G) professional, but can be added to the other five models
BVC basic
- The unpowered version, suitable for use with a VACUULAN™ vacuum network, central vacuum system or shared pump
- Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
- Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl
BVC control
- The BVC control features a powerful whisper-quiet VACUUBRAND™ chemistry-design vacuum pump, eliminating the need for central vacuum supply
- Potentially biohazardous materials can be contained in a local area
- The pump is turned on and off automatically on demand when using the VacuuHandControl, which further reduces the noise level
- An easy to clean touch panel adjusts and indicates the vacuum level switchpoint
BVC professional
- Includes an external liquid level sensor, which operates without contacting bottle contents
- Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
- The standard version is supplied with quick connectors
- The G version includes hose barb connections; optional quick connectors are available
Available Accessories:
- Replacement filter: 20638266
- Replacement VHCpro tubing (sold per meter, minimum order of 2 meters): 20636156
- VHCpro hand controller (one supplied with unit): 20688061
- VHCpro stand: 20635680
- Pipette tip ejector for VHCpro: 20635638
- 8-channel adapter with tip ejector for VHCpro: 20635679
- BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC): 20696880
- Quick connect for second user (between VHCpro and bottle): 20635807
- Standard connect for second user: 20699943
- Quick connect (between bottle and pump): 20635808
- Replacement 4 L PP bottle (with cap): 20635810
- Replacement 2 L glass bottle (with cap): 20635809
- 2 L glass bottle adapter: 20635839

Specifications
150 mbar (112 Torr) | |
17.9 lb. | |
2 L Glass Bottle, External Liquid Level Sensor | |
BVC Professional G | |
US | |
16.97 lb. | |
104°F | |
10°C | |
50/60 Hz | |
43 cm | |
40.8 cm | |
Chemistry Diaphragm Pump | |
1.1 bar |
Touchpanel | |
8.1 kg | |
100/120 V, 50/60 Hz | |
45 dBA | |
-10°C to +60°C | |
7.7 kg | |
40°C | |
0.4 to 0.5 CFM | |
16.92 in. | |
16.06 in. | |
7.63 in. | |
IP40 | |
100/120 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.