Learn More
Mascon Medical Mascon Medical™ Vacuum Filter, 0.45μm Fullset of 1000ml, Sterile, 1EA/Pack, 12 Packs/Case
SDP
Supplier:  Mascon Medical FSVF0.451000
Encompass_Preferred
The vacuum filter provided by Mascon Medical™ is a disposable filter consumable for solvent filter supplements. This filter is made of PS material and used with a diaphragm vacuum pump. It can filter large-volume samples. The filtered samples can be directly stored in the collection bottle and are ideal for sterile bases, buffers and reagents filtration. A complete vacuum filter set is composed of a polypropylene cup cover and cup, a connector, a filter membrane and reservoir bottle. Features:•The polypropylene cup has a GL-45 thread and fits most glass and plastic media storage bottles•Meets ISO13485 quality management system standard•Sterilized by irradiation, SAL 10-6•Individually packed•Conform to USP CLASS VI•Nonpyrogenic•DNase/RNase FreeSpecifications:•Membrane Pore Size: 0.45µm•Reservoir Bottle Capacity: 1000mL•Qty. / Case: 12•Size: Φ98*253mm•Material: Polypropylene, Polystyrene, and ABS•Sterile: Yes•Packaging Format: 1 EA/Bag, 12 Bags/Case
Catalog No. NC2458394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.