Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Vacuum Filtration Device Clamp

Securely clamp flasks to avoid spillage and breakage when using vacuum/suction filtration setups with glass frits. Whatman™ Vacuum Filtration Device Clamp includes spring or locking clamps for use with GV 025/0 or GV 100/0 vacuum filter units.

$84.86 - $579.07

Specifications

For Use With (Application) Vacuum/Suction Filtration
Temperature (Metric) Max 200°C
Type Clamp
View More Specs

Products 2
Catalog Number Mfr. No. Description For Use With Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With Price Quantity & Availability  
09-920-135
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10443005
Locking Clamp GV 100/0 vacuum filter device Each for $579.07
Add to cart
 
09-920-130
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10441005
Spring Action Clamp GV 025/0 vacuum filter device Each for $84.86
Add to cart
 
Description

Description

Whatman glass vacuum filtration devices with glass frit filter, rubber stopper connection and membrane filter accessories are chemically resistant to most aqueous, organic, acidic and caustic solutions. They are autoclavable and can be used up to 200°C.

  • GV 025/0/06: Spring action clamp for 60mL vacuum filtration unit
  • GV 100/0/06: Locking clamp with silicone O-ring for 500mL vacuum filtration unit
Specifications

Specifications

Vacuum/Suction Filtration
200°C
Clamp
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit