Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front Left/Right and Rear Left/Right Connection
Shop All Kemtech Products
Description
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.
Hose connection : Back Beam- Both Ends, Front Beam- Both Ends
Specifications
Specifications
| Connection Type | Front L/R, Rear L/R Hose Connection |
| Type | Vacuum/Inert Gas Double Manifold |
| For Use With (Application) | Organic Synthesis |
| Includes | Hollow Glass Stopcocks |
| Material | Borosilicate Glass |
| No. of Ports | 4 |
| Width (Metric) | 375 mm |