Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front Left/Right, Rear/Right Hose Connection
Shop All Kemtech Products
Description
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.
Hose connection : Back Beam-Right Only, Front Beam-Both Ends
Specifications
Specifications
| Connection Type | Front L/R, Rear Right Hose Connection |
| Type | Vacuum/Inert Gas Manifold |
| For Use With (Application) | Organic Synthesis |
| Includes | Hollow Glass Stopcocks |
| Material | Borosilicate Glass |
| No. of Ports | 4 |
| Width (Metric) | 375 mm |