Learn More
Chemglass Life Sciences Vacuum/Inert Manifold, 4 Ports, 365mm Overall Width
SDP
Supplier:  Chemglass Life Sciences CG444002
Encompass_Preferred
Vacuum/Inert Manifold, 4 Ports, 365mm Overall Width. General purpose double bank vacuum manifold with 4 ports for use with vacuum and inert gas. Both tubes are 1/2” OD with serrated hose connections located on the upper left and lower right banks. Ports are 4mm high vacuum stopcocks with serrated hose connections. Ports are spaced 85mm center to center. All hose connections have a 10mm small serration and a 11mm large serration.
Catalog No. CG444002
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.