Learn More
BINDER™ Vacuum Module with Vacuum Pump VP 1
SDP
Pump chamber for installation of vacuum pumps, with switchable power socket. Including chemical membrane pump with a nominal air flow of 2.0 m3/hour, final pressure 7 mbar and connection kit VD VP1-VP2.
Supplier:  BINDER™ 80122626
 View more versions of this product
Catalog No. 11-103-192
Only null left
Add to Cart
Description
Description
  • Temperature range:
  • +9°C above ambient temperature to +220 °C
  • Controller with digital display of pressure and temperature
  • Program-controlled drying monitoring with automatic ventilation at end of process
  • Internal data logger, measured values can be read out in open format via USB
  • 1 aluminum expansion rack, can be custom-positioned
  • Inert gas connection
  • Universal access port DN 16 - for the introduction of external measuring devices
  • Shatterproof, spring-mounted safety glass panel
  • Large viewing window with option of interior lighting
Specifications
Specifications
1.8 to 3.4
581 mm
220°C
942 mm
743 mm
50/60 Hz
70 kg
BINDER units VD115UL-120V
581 mm
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.