Learn More
BINDER™ Vacuum Module with Vacuum Pump VP 2
SDP
Catalog No. 11103193 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With:
BINDER units VD023UL-120V
BINDER units VD056UL-120V
BINDER units VD115UL-120V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11-103-193 BINDER units VD115UL-120V
11-103-198 BINDER units VD023UL-120V
11-103-200 BINDER units VD056UL-120V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-103-193 Supplier BINDER™ Supplier No. 80122627
Add to Cart
Add to Cart
Request a Quote
Vacuum module with vacuum pump VP 2 - 120 V / 50-60 Hz option model, Pump chamber for installation of vacuum pumps, with switchable power socket.
  • Temperature range:
  • +9°C above ambient temperature to +110 °C
  • Controller with digital display of pressure and temperature
  • Program-controlled drying monitoring with automatic ventilation at end of process
  • Internal data logger, measured values can be read out in open format via USB
  • 1 aluminum expansion rack, can be custom-positioned
  • Inert gas connection
  • Universal access port DN 16 - for the introduction of external measuring devices
  • Shatterproof, spring-mounted safety glass panel
  • Large viewing window with option of interior lighting
Specifications
Electrical Requirements 50/60 Hz
Dimensions (L x W x H) 581 x 743 x 942 mm
Temperature (Metric) Max 110°C
For Use With BINDER units VD115UL-120V
Voltage 120 V