Learn More
Sheldon Manufacturing Inc VACUUM OVEN 1.67CF 115V
SDP
Supplier:  Sheldon Manufacturing Inc SVAC2
Encompass_Preferred
VACUUM OVEN, PROGRAMMABLE DIGITAL MODEL, 1.67 CU FT, 115V This programmable oven can host up to 40 ramp-and-soak temperature control steps and has a digital vacuum display. SHEL LAB vacuum ovens provide exceptional capabilities for fast and gentle drying of heat sensitive materials. They prevent residue accumulation on products and also allow safe drying of materials at lower temperatures than typically required when using convection ovens. Please note that this vacuum oven does not include a pump or related accessories. Contact your SHEL LAB distributor for assistance with choosing the ideal pump and accessories for any application. More information available at www.shellab.com
Catalog No. NC1198553
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.