Learn More
BUCHI Vacuum Pump Silencer
SDP
Catalog No. 05405064 Shop All BUCHI Products
Change view
Click to view available options
Material:
PBT
Catalog No. Material
05-405-064 PBT
1 options
Catalog No. 05-405-064 Supplier BUCHI Supplier No. 11062291
Silencer attaches at the back of the vacuum pump to absorb sound.
  • Used to absorb sound from the vacuum pump
  • Compatible with Vacuum Pumps V-700, V-710, V-100, V-300 and V-600
Specifications
For Use With (Application) Reducing Noise From Vacuum Pumps
For Use With Vacuum Pumps: V-100, V-300, V-600, V-700, V-710
Material PBT
Type Silencer