Learn More
Binder Connection Kit VD VP1-VP2
SDP
For vacuum pump VP1 and VP2.
Supplier:  Binder 80122613
Catalog No. 11-103-189
Only null left
Add to Cart
Description
Description
  • Consisting of plastic hose 1.9 m long, clamping ring, universal centering ring and small flange adapter 1/2 in. in diameter (DN 16) to hose nozzle 3/8 in. in diameter (DN 10).
Specifications
Specifications
Vacuum Pump VP1 and VP2
IP40
Pump Connection Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.