Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Vacuum Set with Level Sense Assembly
GREENER_CHOICE

Catalog No. BT7100543S Shop All Agilent Technologies Products
Change view
Click to view available options
Capacity (Metric):
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
BT7100543S 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BT7100543S Supplier Agilent BioTek Supplier No. 7100543S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vacuum set for use with the 405 TS, 405 LS, ELx405 Select Deep Well, EL406 and 406 FX.

  • Includes 4 or 10 L bottles
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4 L
For Use With (Equipment) 405 TS Microplate Washer, 405 LS Microplate Washer, ELx 405 Select Deep Well Microplate Washer, EL406 Microplate Washer, 406 FX Microplate Washer
Product Type Vacuum Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.