Learn More
Instech Rat VAH tether assembly, low dead volume 017 PU tubing for ABS, 10.5in, sterile
SDP
Supplier:  Instech VAH95T/ABS
Encompass_Preferred
Mating tether for Vascular Access Harness™ for rats. 015 silicone in fluid path. Low dead volume for use with automated blood samplers.
Catalog No. 50-812-48
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.