Description
- Core2Clean Plus systems can be configured to operate as three different systems: a sprayer, a wet mop, or a fogger
- Design of the Core2Clean Plus system allows for repeatability between operators and areas, making it easier for companies to write SOP’s for different class areas
- Veltek provides an Operator’s Manual with instructions for all three system uses, and videos of select processes
Specifications
Specifications
| Product Type | Mop Head |
| Length (English) | 7 in. |
| Sterility | Sterile |
| Resistant To | Abrasion |
| For Use With (Equipment) | Core2Clean VertiKlean System |
| For Use With (Application) | For General Cleaning and Wipe down of Walls and Ceilings |
| Material | Polyester, Foam |