Learn More
VAI Core2Clean Plus Systems
Catalog No. 19546804 Shop All VAI Products
Change view
Click to view available options
Product Type:
Floor Cover
Mop Frame
Mop Head
Length (English):
7 in.
12 in.
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (English) Sterility
19-546-804 Mop Head 12 in. Non-sterile
19-546-790 Mop Head 12 in. Sterile
19-546-792 Floor Cover 7 in. Sterile
19-546-793 Floor Cover 12 in. Non-sterile
19-546-794 Floor Cover 12 in. Sterile
19-546-791 Floor Cover 7 in. Non-sterile
19-546-795 Mop Frame 12 in. Non-sterile
19-546-803 Mop Head 7 in. Sterile
19-546-802 Mop Head 7 in. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-546-804 Supplier VAI Supplier No. C2C1009
The Core2Clean™ Plus Systems were created by Veltek Associates, Inc., to provide specialized cleaning equipment for use in controlled environments.
  • Core2Clean Plus systems can be configured to operate as three different systems: a sprayer, a wet mop, or a fogger
  • Design of the Core2Clean Plus system allows for repeatability between operators and areas, making it easier for companies to write SOP’s for different class areas
  • Veltek provides an Operator’s Manual with instructions for all three system uses, and videos of select processes
Specifications
Product Type Mop Head
Length (English) 12 in.
Sterility Non-sterile
Resistant To Abrasion
For Use With (Equipment) Core2Clean VertiKlean System
For Use With (Application) For General Cleaning and Wipe down of Walls and Ceilings
Material Polyester, Foam