Learn More
VAI DECON-CLEAN Residue Cleaner
Catalog No. 19547013 Shop All VAI Products
Change view
Click to view available options
Volume (English):
4 oz.
11 oz.
16 oz.
1 gal.
Format:
Aerosol Spray
Concentrate
SimpleMix
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Format Sterility
19-547-013 1 gal. Concentrate Non-sterile
19-547-015 4 oz. Concentrate Sterile
19-547-271 11 oz. Aerosol Spray Sterile
19-547-018 16 oz. SimpleMix Sterile
19-547-014 1 gal. Concentrate Sterile
19-547-016 1 gal. SimpleMix Sterile
19-546-500 1 gal. SimpleMix Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-547-013 Supplier VAI Supplier No. DC01
Add to Cart
Add to Cart
DECON-CLEAN Residue Cleaner is designed to remove residues from disinfecting agents.
  • Ideal for washable, nonporous environmental surfaces
  • Use on walls, ceilings, floors, and stainless items
  • Filtered at 0.2 Microns
  • Gamma irradiated
Order Info
Double-bagged packaging. Delivered with lot specific Certificate of Analysis.
Compliance
USP tested and delivered with a lot-specific certificate of irradiation and certificate of analysis. Also comes with an LAL testing analysis and sterility report.
Specifications
Sterility Non-sterile
Certifications/Compliance USP, ISO
Filtered Filtered
Format Concentrate
For Use With (Application) Removes residues from sanitizers, disinfectants and sporicides, used for walls, ceilings, floors, stainless items, countertops, aseptic connections etc.
Includes Certificate of analysis, certificate of irradiation and certificate of sterility
Volume (English) 1 gal.
Volume (Metric) 3.8 L