Learn More
Description
DECON-CLEAN Residue Cleaner is designed to remove residues from disinfecting agents.
- Ideal for washable, nonporous environmental surfaces
- Use on walls, ceilings, floors, and stainless items
- Filtered at 0.2 Microns
- Gamma irradiated
Order Info
Double-bagged packaging. Delivered with lot specific Certificate of Analysis.
Compliance
USP tested and delivered with a lot-specific certificate of irradiation and certificate of analysis. Also comes with an LAL testing analysis and sterility report.
Specifications
Specifications
| Sterility | Sterile |
| Certifications/Compliance | USP, ISO |
| Format | Concentrate |
| For Use With (Application) | Removes residues from sanitizers, disinfectants and sporicides, used for walls, ceilings, floors, stainless items, countertops, aseptic connections etc. |
| Includes | Certificate of analysis, certificate of irradiation and certificate of sterility |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.8 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.