Learn More
Description
Includes
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
A detergent suitable for controlled areas which is effective in 10 minutes at 20°C in hard water up to 400ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
- Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface
- Filtered at 0.2μm and gamma irradiated
- Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces
- Completely traceable and validated for sterility and shelf life
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Specifications
| Form | Concentrate |
| Sterility | Sterile |
| For Use With (Application) | Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries |
| Volume (English) | 1 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.