Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-QUAT 100 Quaternary Ammonium Solution

Catalog No. 19547054 Shop All VAI Products
Change view
Click to view available options
Volume (English):
8 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
Format:
Concentrate
SimpleMix
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Format
19-547-054 1 gal. Sterile SimpleMix
19-546-850 1 gal. Non-sterile Concentrate
19-546-851 1 gal. Sterile Concentrate
19-547-055 1 gal. Non-sterile SimpleMix
19-547-056 16 oz. Sterile SimpleMix
19-546-853 8 oz. Sterile Concentrate
19-547-057 16 oz. Non-sterile SimpleMix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-547-054 Supplier VAI Supplier No. DQ100042Z
Add to Cart
Add to Cart

DECON-QUAT 100 Quaternary Ammonium Solution for use on hard, nonporous environmental surfaces in medical and nonmedical facilities.

  • Effective as a one-step disinfectant, virucide, deodorizer, sanitizer and fungicide
  • For use on stainless steel, porcelain, glass and chrome
  • Filled in ISO 5 (Grade A/B, Class 100)
  • Filtered at 0.2 micron
  • Completely validated for sterility and shelf life
  • Hard, inanimate surfaces in filling and gowning rooms, general manufacturing areas, machinery tables, counter tops, and laminar flow benches, floors and wall

Specifications

Product Type Quaternary Ammonium Solution
Sterility Sterile
Certifications/Compliance USP, ISO
Composition Active ingredients of 5% Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chloride and 5% Akly (68% C12, 32% C14) Dimethyl Ethylbenzyl Ammonium Chloride and 90% inert ingredients
Dilution 2:128
Filtered Filtered
Format SimpleMix
For Use With (Application) Used as a disinfectant on inanimate, hard, non porous environmental surfaces such as walls, ceilings, floors, countertops, general manufacturing areas
Includes Certificate of analysis, certificate of irradiation and certificate of sterility
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.