Learn More
Description
Includes
Lot-specific Certificate of Analysis, LAL testing analysis and sterility report
DECON-SPORE 200 Plus Disinfectant Solution is designed for the sterilization of manufacturing, packaging and filling equipment in aseptic processes.
- Decon-Spore 200 Plus is a Sterilant-Virucide-Disinfectant-Sanitizer-Booster for alkaline detergents
- Effective sanitizer on inanimate, non-porous, non-food contact surfaces
- Sterile pharmaceutical cleanroom formula
- Phosphate free
- Peracetic acid and hydrogen peroxide solution filtered at 0.2 μm
- Disinfects as it cleans in one operation
- Broad-spectrum hard surface disinfectant effective against influenza A viruses
- Filled in ISO 5 (Grade A/B, former Class 100)
- Double-bagged packaged
- Available in several container types (unit dose, concentrate and SimpleMix) and sizes both sterile and nonsterile
- Completely traceable and validated for sterility and shelf life
Simple Mix System
- Ready to use package
- Holds concentrate in small chamber and WFI quality water in larger chamber
- Simply press top of small chamber to release concentrate into water and agitate to mix
- Eliminates risk of operator error when measuring appropriate amount of concentrate and gives fresh solution every time
Order Info
Suggested minimum surface exposure time for 5% dilution based on the temperature of the solution: 20°C for 6 hours, 50°C for 20 minutes, or 80°C for 5 minutes (68°, 122° or 176°F). In the presence of 5% blood serum and soap film on a nonporous surface, suggested surface exposure time of 10 minutes at 20°C.
Specifications
Specifications
| Sterility | Sterile |
| Certifications/Compliance | USP, AOAC, ISO |
| Composition | Peracetic Acid and Hydrogen Peroxide solution |
| Concentration | 5% for sterilization, 0.3% for disinfection |
| Dilution | 3 oz./gal. |
| Filtered | Filtered |
| Format | SimpleMix (3% Disinfectant/Sporicidal Dose) |
| For Use With (Application) | Designed for the sterilization of manufacturing, packaging and filling equipment in aseptic processes, to disinfect floors, walls and other hard non porous surfaces |
| Includes | Lot specific certificate of analysis and certificate of sterility |
| Packaging | Attached Trigger Spray |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.