Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI Process2Clean 6 Chlorinated Alkaline Cleaner

Catalog No. 19546911 Shop All VAI Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
55 gal.
Sterility:
Non-sterile
Sterile
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility
19-546-911 55 gal. Sterile
19-546-906 1 gal. Non-sterile
19-546-907 1 gal. Sterile
19-546-908 30 gal. Non-sterile
19-546-909 30 gal. Sterile
19-546-910 55 gal. Non-sterile
19-546-912 5 gal. Non-sterile
19-546-913 5 gal. Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-546-911 Supplier VAI Supplier No. PC655G02
Add to Cart
Add to Cart

Process2Clean 6 is a chlorinated alkaline cleaning agent. P2C 6 contains buffering agents to maintain substrate integrity. P2C 6 is capable of removing highly proteinaceous soils, oils, waxes, creams, and polymers.

  • Characteristic: Alkaline
  • Clarity: Clear
  • Appearance: Yellow
  • pH (1% solution) : 11.0 – 13.0
  • Conductivity (1% solution): 6.50 – 8.50 mS

Specifications

Form Liquid
pH 12.6
Sterility Sterile
Appearance Clear
Content And Storage 89.6°C to 176°C
For Use With (Application) For low energy agitate immersion systems, pressure spray, or manual washing
Volume (English) 55 gal.
Volume (Metric) 208.2 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.