Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Stainless Steel Compressed Air Atrium, Complete Unit

Catalog No. p-4910915 Shop All VAI Products
Change view
Click to view available options
For Use With (Application):
18 mL Filled Agar Plates
25 mL Filled Agar Plates
32 mL Filled Agar Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
19-547-096 18 mL Filled Agar Plates
19-547-097 25 mL Filled Agar Plates
19-547-098 32 mL Filled Agar Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-547-096 Supplier VAI Supplier No. SMA316CA18
Only null left
Add to Cart
Add to Cart
Request a Quote

An SMA Compressed Air/Gas Atrium is a 316 Stainless Steel capture device that is connected to a compressed air source. It has an extension tube that attaches to 1/4 inch Hytrel™ tubing.

  • Exhaust air is released from underneath the Atrium without affecting the sample in progress
  • Compact size allows them to be located near filling processes where space is limited
  • Available as a complete top and bottom assembly
  • Vents from the bottom which eliminates the need for additional vacuum connection ports or tubing
  • Connection: 0.25 in. Hytrel™ Tubing

Specifications

Product Type SMA Compressed Air/Gas Atrium Complete Assembly
Material Stainless Steel
For Use With (Application) 18 mL Filled Agar Plates
Airflow Rate 1 to 1.5 cfm
Overall Height (Metric) 3.81 cm
Overall Height (English) 1.5 in.
Diameter (Metric) Outer 11.43 cm
Diameter (English) Outer 4.5 in.
Exposure Time 50 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.