Description
- Exhaust air is released from underneath the Atrium without affecting the sample in progress
- Compact size allows them to be located near filling processes where space is limited
- Available as a complete top and bottom assembly
- Vents from the bottom which eliminates the need for additional vacuum connection ports or tubing
- Connection: 0.25 in. Hytrel™ Tubing
Specifications
Specifications
| Product Type | SMA Compressed Air/Gas Atrium Complete Assembly |
| For Use With (Application) | 25 mL Filled Agar Plates |