Learn More
Description
- Specifically formulated as a sterile pharmaceutical cleanroom formula
- Lubricates for continuous and effortless manufacturing
- Designed for all washable environmental surfaces
- Filled in an ISO 5 (Grade A/B, Former Class 100) operation
- Filtered at 0.2 microns
- Components are air washed with 0.2 micron filtered air before assembly
Specifications
Specifications
| pH | 9 |
| Sterility | Sterile |
| Certifications/Compliance | Lot specific Certificate of Analysis, Certificate of Sterility, and Certificate of Irradiation |
| Filtered | Filtered |
| Packaging | Bottle |
| Volume (English) | 1 gal. |
| Volume (Metric) | 3.8 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.