Learn More
USP Purified Steri-Water, VAI
Catalog No. 19546944 Shop All VAI Products
Change view
Click to view available options
Volume (English):
16 oz.
1 gal.
2 gal.
5 gal.
Packaging:
Bottle
Bottle with Unattached Trigger
Gallon Drum, Unattached Faucet
Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Packaging
19-546-944 5 gal. Gallon Drum, Unattached Faucet
19-546-942 16 oz. Bottle with Unattached Trigger
19-546-941 1 gal. Bottle
19-546-943 2 gal. Bottle
19-547-288 - Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-546-944 Supplier VAI Supplier No. STWA5G
Add to Cart
Add to Cart
USP Purified Steri-Water designed for use in the pharmaceutical, biotechnology, and medical device industries for chemical formulations, cleaning, rinsing and many other applications.
  • Specifically formulated as a sterile pharmaceutical cleanroom formula
  • Lubricates for continuous and effortless manufacturing
  • Designed for all washable environmental surfaces
  • Filled in an ISO 5 (Grade A/B, Former Class 100) operation
  • Filtered at 0.2 microns
  • Components are air washed with 0.2 micron filtered air before assembly
Specifications
Sterility Sterile
Certifications/Compliance Lot specific Certificate of Analysis, Certificate of Sterility, and Certificate of Irradiation
Filtered Filtered
Packaging Gallon Drum, Unattached Faucet
Volume (English) 5 gal.
Volume (Metric) 18.9 L