Learn More
Description
- Filled in an ISO 5 (Grade A/B, Former Class 100) operation
- Filtered at 0.2 microns
- Components are air washed with 0.2 micron filtered air before assembly
- Lot sterility tested according to current USP compendium
- Lot tested for endotoxins
Specifications
Specifications
| Format | Liquid |
| Quantity | 2 gal. (7.6 L) |
Safety and Handling
| ShelfLife | 2 Years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
